5-Bromo-1-pentene, 95%
5-Bromo-1-pentene, 95%
5-Bromo-1-pentene, 95%
Acros Organics

5-Bromo-1-pentene, 95%

CAS: 1119-51-3 | C5H9Br | 149.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
2.5 g
10 g
Packaging:
Glass bottle
Catalog number 295170500
also known as 29517
HSN 29033919
Price (INR)
37,590.00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
37,590.00
Each
Chemical Identifiers
CAS1119-51-3
IUPAC Name5-bromopent-1-ene
Molecular FormulaC5H9Br
InChI KeyLPNANKDXVBMDKE-UHFFFAOYSA-N
SMILESBrCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
AppearanceMay darken on storage
Infrared spectrumConforms
GC>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only