Lithium acetate, 99+%, for analysis, anhydrous
Lithium acetate, 99+%, for analysis, anhydrous
Lithium acetate, 99+%, for analysis, anhydrous
Lithium acetate, 99+%, for analysis, anhydrous
Acros Organics

Lithium acetate, 99+%, for analysis, anhydrous

CAS: 546-89-4 | C2H3LiO2 | 65.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 297111000
HSN 29152990
Price (INR)
16,207.00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
16,207.00
Each
Add to cart
Chemical Identifiers
CAS546-89-4
IUPAC Namelithium(1+) acetate
Molecular FormulaC2H3LiO2
InChI KeyXIXADJRWDQXREU-UHFFFAOYSA-M
SMILES[Li+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO499.0 to 101.0 %
Melting point280°C to 285°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only