L-tert-Leucine, 99%
L-tert-Leucine, 99%
L-tert-Leucine, 99%
Acros Organics

L-tert-Leucine, 99%

CAS: 20859-02-3 | C6H13NO2 | 131.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
297780250
also known as 29778
25 gGlass Bottle
Catalog number 297780250
also known as 29778
HSN 29224990
Price (INR)
42,585.00
Each
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS20859-02-3
IUPAC Name(2S)-2-amino-3,3-dimethylbutanoic acid
Molecular FormulaC6H13NO2
InChI KeyNPDBDJFLKKQMCM-SGAVLPGINA-N
SMILESCC(C)(C)[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Heavy metals (as Pb)=<10 ppm
Iron (Fe)=<10 ppm
Infrared spectrumConforms
Loss on drying=<0.2 % (105°C, 3 hrs)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only