Product Image
Thermo Scientific™

Avidin Protein, HRP

Avidin for Western Blot, IHC, ELISARead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
299945 mg
211232 mg
Catalog number 29994
Price (INR)
60,281.00
Online offer
Ends: 31-Mar-2026
100,467.00
Save 40,186.00 (40%)
Each
Quantity:
5 mg
Request bulk or custom format
Price (INR)
60,281.00
Online offer
Ends: 31-Mar-2026
100,467.00
Save 40,186.00 (40%)
Each
Avidin for Western Blot, IHC, ELISA
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeHRP (Horseradish Peroxidase)
Quantity5 mg
Target MoleculeBiotin
EnzymeHRP (Horseradish Peroxidase)
For Use With (Application)Western Blotting, ELISA, Immunohistochemistry
FormSolid
Product LinePierce
SpecificityBiotin
Unit SizeEach
Contents & Storage
Store at -20°C. Shipped at ambient temperature.