Bis(cyclopentadienyl)dimethyltitanium, 5 wt.% in toluene, AcroSeal™
Bis(cyclopentadienyl)dimethyltitanium, 5 wt.% in toluene, AcroSeal™
Bis(cyclopentadienyl)dimethyltitanium, 5 wt.% in toluene, AcroSeal™
Bis(cyclopentadienyl)dimethyltitanium, 5 wt.% in toluene, AcroSeal™
Acros Organics

Bis(cyclopentadienyl)dimethyltitanium, 5 wt.% in toluene, AcroSeal™

CAS: 1271-66-5 | C12H20Ti | 212.159 g/mol
Have Questions?
Catalog NumberQuantityPackaging
301761000
also known as 30176
100 mLAcroSeal™ Glass Bottle
Catalog number 301761000
also known as 30176
HSN 29319090
Price (INR)
41,780.00
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (INR)
41,780.00
Each
Chemical Identifiers
CAS1271-66-5
IUPAC Namebis(cyclopenta-1,3-diene) bis(methane) titanium
Molecular FormulaC12H20Ti
InChI KeyCRKLPNOLYBORCP-UHFFFAOYSA-N
SMILESC.C.[Ti].C1C=CC=C1.C1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to dark red
NMR (1H-NMR)Consistent with structure
Appearance (Form)Solution
Strength5 % (approximately)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only