Search
Search

| Catalog Number | Color | Holds | Array |
|---|---|---|---|
| 30190 | Blue | Tubes (1.5 mL) | 8 x 12 |
| 30180BI | Red | Tubes (10 to 13 mm) | 6 x 12 |
| 30184 | White | Tubes (17 to 20 mm) | 4 x 10 |
| 30187 | Blue | Tubes (21 to 25 mm) | 4 x 4 |
Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.
Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.