beta-Tetralone, 95%
beta-Tetralone, 95%
beta-Tetralone, 95%
Acros Organics

beta-Tetralone, 95%

CAS: 530-93-8 | C10H10O | 146.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 302830050
also known as 30283
HSN 29143990
Price (INR)
7,220.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
7,220.00
Each
Chemical Identifiers
CAS530-93-8
IUPAC Name1,2,3,4-tetrahydronaphthalen-2-one
Molecular FormulaC10H10O
InChI KeyKCKZIWSINLBROE-UHFFFAOYSA-N
SMILESO=C1CCC2=CC=CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to green
Infrared spectrumConforms
Appearance (Form)Liquid or low melting solid
Refractive index1.5550 to 1.5650 (20°C, 589 nm)
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only