Sodium metatungstate, 2.82 density solution in water, stabilized
Sodium metatungstate, 2.82 density solution in water, stabilized
Sodium metatungstate, 2.82 density solution in water, stabilized
Acros Organics

Sodium metatungstate, 2.82 density solution in water, stabilized

CAS: 12141-67-2 | H2Na6O40W12
Have Questions?
Catalog NumberQuantityPackaging
304100500
also known as 30410
50 mLGlass bottle
Catalog number 304100500
also known as 30410
HSN 28418090
Price (INR)
24,500.00
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
24,500.00
Each
Chemical Identifiers
CAS12141-67-2
IUPAC Namehexasodium tris(dioxotungstenbis(olate)) nonakis(trioxotungsten)
Molecular FormulaNa6O39W12
InChI KeyAZCSOJKJFMWYCX-UHFFFAOYSA-N
SMILES[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
StabilizerTypically 0.02 % Na2Cr2O7 salt
Appearance (Color)Clear yellow to light green
Assay>=67 % (WO3)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only