4-Bromophenol, 97%
4-Bromophenol, 97%
4-Bromophenol, 97%
Acros Organics

4-Bromophenol, 97%

CAS: 106-41-2 | C6H5BrO | 173.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 304415000
also known as 30441
HSN 29081900
Price (INR)
24,500.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
24,500.00
Each
Chemical Identifiers
CAS106-41-2
IUPAC Name4-bromophenol
Molecular FormulaC6H5BrO
InChI KeyGZFGOTFRPZRKDS-UHFFFAOYSA-N
SMILESOC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Infrared spectrumConforms
Appearance (Color)White to pink to light brown
Appearance (Form)Crystalline powder or crystals or solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only