2-Methylbutyraldehyde, 95%
2-Methylbutyraldehyde, 95%
2-Methylbutyraldehyde, 95%
Acros Organics

2-Methylbutyraldehyde, 95%

CAS: 96-17-3 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 304470500
HSN 29122990
Price (INR)
7,409.00
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
7,409.00
Each
Chemical Identifiers
CAS96-17-3
IUPAC Name2-methylbutanal
Molecular FormulaC5H10O
InChI KeyBYGQBDHUGHBGMD-UHFFFAOYNA-N
SMILESCCC(C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=94 %
Refractive index1.3890 to 1.3950 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only