2-Methylene-1,3-propanediol, 97%
2-Methylene-1,3-propanediol, 97%
2-Methylene-1,3-propanediol, 97%
Acros Organics

2-Methylene-1,3-propanediol, 97%

CAS: 3513-81-3 | C4H8O2 | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 308590050
also known as 30859
HSN 29053990
Price (INR)
19,860.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
19,860.00
Each
Chemical Identifiers
CAS3513-81-3
IUPAC Name2-methylidenepropane-1,3-diol
Molecular FormulaC4H8O2
InChI KeyJFFYKITVXPZLQS-UHFFFAOYSA-N
SMILESOCC(=C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Viscous liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4730 to 1.4750 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only