Nonafluoropentanoic acid, 97%
Nonafluoropentanoic acid, 97%
Nonafluoropentanoic acid, 97%
Acros Organics

Nonafluoropentanoic acid, 97%

CAS: 2706-90-3 | C5HF9O2 | 264.05 g/mol
Have Questions?
Catalog NumberQuantityPackaging
309290250
also known as 30929
25 mLGlass bottle
Catalog number 309290250
also known as 30929
HSN 29159090
Price (INR)
51,556.00
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
51,556.00
Each
Chemical Identifiers
CAS2706-90-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Titration with NaOH>=96.0 %
Appearance (Color)Clear light yellow to light brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only