Pierce™ High Sensitivity NeutrAvidin™-HRP
Thermo Scientific™

Pierce™ High Sensitivity NeutrAvidin™-HRP

Thermo Scientific Pierce High Sensitivity NeutrAvidin HRP Conjugate is a specially prepared peroxidase-conjugated form of avidin biotin-binding protein that providesRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
310325 mL
310300.5 mL
Catalog number 31032
Price (INR)
-
Quantity:
5 mL
Request bulk or custom format

Thermo Scientific Pierce High Sensitivity NeutrAvidin HRP Conjugate is a specially prepared peroxidase-conjugated form of avidin biotin-binding protein that provides signal amplification like poly-HRP and exceptional storage stability.

Features of High Sensitivity NeutrAvidin HRP Conjugate:

  • NeutrAvidin Protein—a specially deglycosylated form of avidin that provides highly specific, low-background binding of biotinylated antibodies in many applications.
  • High sensitivity—detect low levels of target without background; obtain high signal-to-noise ratios
  • Cost effective—use less conjugate in Western blotting and ELISA applications and still obtain excellent results
  • Flexible—compatible with typical chemiluminescent, fluorescent and colorimetric peroxidase substrates
  • Convenient—ready-to-use stabilized liquid format means there is no waiting for thawing and no need to aliquot

This specially-manufactured variety of HRP-conjugated avidin protein meets the demands of today's scientists for more sensitive detection in ELISA and Western blotting applications. The conjugate is suitable for use with chemiluminescent, chemifluorescent or colorimetric substrates. Each High Sensitivity HRP conjugate is packaged in an easy-to-use stabilized solution that enables convenient storage at 4°C for at least one year.

Our High Sensitivity HRP Conjugates outperform the competition on target detection, signal intensity and signal-to-noise ratios. For example, experiments indicate that the High Sensitivity Streptavidin-HRP can be diluted eight-to 10-fold more than traditional HRP conjugates and still produce similar results.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeHRP (Horseradish Peroxidase)
Quantity5 mL
Target MoleculeBiotin
Detection MethodColorimetric, Chemiluminescence, Chemifluorescence
EnzymeHRP (Horseradish Peroxidase)
For Use With (Application)Western Blotting, ELISA
FormLiquid
Product LinePierce
SpecificityBiotin
Unit SizeEach
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2°C to 8°C.