Product Image
Thermo Scientific™
Orion™ 3106 Chemical Oxygen Demand Analyzer Reagent and Standards
Designed and packaged for our Thermo Scientific™ Orion™ 3106 COD analyzer. Reagents are prepackaged as a kit to provide all reagents for 30 days of operation. The standards are available in individual bottles to allow the appropriate solutions to be used with the range of interest.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
310610003106 COD standard, 1000ppm
3106RE3106 COD Reagent
31060403106 COD standard, 40ppm
31061603106 COD standard, 160ppm
Catalog number 31061000
Price (INR)
-
Description:
3106 COD standard, 1000ppm
Designed and packaged for our Thermo Scientific™ Orion™ 3106 COD analyzer. Reagents are prepackaged as a kit to provide all reagents for 30 days of operation. The standards are available in individual bottles to allow the appropriate solutions to be used with the range of interest.
Warranty: Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
Specifications
Description3106 COD standard, 1000ppm
TypeStandards
Unit SizeEach