Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure: Bulk Pack
Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure: Bulk Pack
Actual product may vary
Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure: Bulk Pack
Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure: Bulk Pack
Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure: Bulk Pack

Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure: Bulk Pack

Package, ship and store a variety of reagents using these round, narrow-mouth LDPE bottles with closure.
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
312003-0016500 mL28 mm
Catalog number 312003-0016
HSN 39233090
Price (INR)
37,899.00
Case
Add to cart
Capacity (Metric):
500 mL
Closure Size:
28 mm
Price (INR)
37,899.00
Case
Add to cart

Package, ship and store a variety of reagents using Thermo Scientific™ Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure, constructed from natural low-density polyethylene.

Key Features
  • Durable and clear
  • Uniform walls that resist splitting and puncturing
  • Leakproof† when used with Nalgene closures
  • Ideal for trace metal analysis — LDPE is low in trace metal content

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionNarrow-Mouth Boston Round Bottle
Diameter (Metric) Inner Neck21 mm
MaterialLDPE
No. per Case125
No. per Pack125
SterilityNon-sterile
Closure ColorNatural
Neck Finish28 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)170 mm
Product TypeNarrow Mouth Bottle
Width (English)2.86 in.
Height (English)6.71 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size28 mm
Diameter (Metric)73 mm
Closure TypeYes (Bulk Packed, Unassembled)
Through the Closure DispensingNo
Unit SizeCase