Nalgene™ Lined HDPE Serum Vial Closure for Continuous-Thread Vials: Nonsterile
Actual product may vary
Nalgene™ Lined HDPE Serum Vial Closure for Continuous-Thread Vials: Nonsterile
Nalgene™ Lined HDPE Serum Vial Closure for Continuous-Thread Vials: Nonsterile
Nalgene™ Lined HDPE Serum Vial Closure for Continuous-Thread Vials: Nonsterile

Nalgene™ Lined HDPE Serum Vial Closure for Continuous-Thread Vials: Nonsterile

Eliminate the need for a rubber stopper while maintaining a leakproof seal with these lined, HDPE serum vial closures, for use with continuous thread PETG serum vials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
312158-0021White
312158-0022Black
Catalog number 312158-0021
HSN 39235090
Price (INR)
139,415.00
Case
Color:
White
Price (INR)
139,415.00
Case

Eliminate the need for a rubber stopper, yet maintain a leakproof† seal with Thermo Scientific™ Nalgene™ Lined HDPE Serum Vial Closures for Continuous-Thread Vials. These nonsterile, high density polyethylene closures are for use with continuous-thread PETG serum vials.

  • Three-ply co-extruded liner (LDPE core between two facings of solid HDPE)
  • For use with nonsterile Nalgene PETG serum vials

Ordering Information:

Serum vial closures are bulk packed 2,000 per case.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Closure ColorWhite
DescriptionSerum vial closures
Material (Liner)Three-ply co-extruded liner (LDPE core between two facing of solid HDPE)
ColorWhite
Diameter (Metric) Outer22.1 mm
MaterialHDPE
SterilityNon-sterile
No. per Case2000
No. per Pack2000
Product LineNalgene
Closure MaterialHDPE
Closure TypeScrew Cap
Quantity2000 Cs.
Closure Size20-415
For Use With (Equipment)Nonsterile Nalgene PETG Serum Vials
Certifications/ComplianceISO 13485:2003
Height (Metric)14.2 mm
Product TypeSerum Vial Closure
Unit SizeCase