Tin(II) trifluoromethanesulfonate, 98%
Tin(II) trifluoromethanesulfonate, 98%
Tin(II) trifluoromethanesulfonate, 98%
Acros Organics

Tin(II) trifluoromethanesulfonate, 98%

CAS: 62086-04-8 | C2F6O6S2Sn | 416.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Plastic bottle
Catalog number 312530050
also known as 31253
HSN 29319090
Price (INR)
22,322.00
Each
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
22,322.00
Each
Chemical Identifiers
CAS62086-04-8
IUPAC Namebis(trifluoromethanesulfonic acid) tin
Molecular FormulaC2H2F6O6S2Sn
InChI KeyHKYRUGOZNFKLHI-UHFFFAOYSA-N
SMILES[Sn].OS(=O)(=O)C(F)(F)F.OS(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
Assay>=97.5 % (ex Sn)
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only