Septa
Actual product may vary
Septa
Septa
Septa
Septa
Septa
Septa
A wide range to cover most applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) Inner
3130323311 mm
3130321017 mm
31313225
also known as 31313225S
12.7 mm
Catalog number 31303233
HSN 40169990
Price (INR)
19,290.00
Each
Diameter (Metric) Inner:
11 mm
Price (INR)
19,290.00
Each
A wide range to cover most applications
  • Variety of materials and temperature limits for all applications
  • For use in Thermo Scientific TRACE™ and FOCUS™ GCs and instruments from other manufacturers
  • Contaminant-free in individual blister packs
Specifications
For Use With (Equipment)GC Instruments
MaterialSilicone
TypeLow Bleed Septum
Diameter (Metric) Inner11 mm
QuantityPack of 50
Unit SizeEach