3,5-Dichlorophenyl isocyanate, 96%
3,5-Dichlorophenyl isocyanate, 96%
3,5-Dichlorophenyl isocyanate, 96%
Acros Organics

3,5-Dichlorophenyl isocyanate, 96%

CAS: 34893-92-0 | C7H3Cl2NO | 188.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 314140250
also known as 31414
HSN 29291020
Price (INR)
23,293.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
23,293.00
Each
Chemical Identifiers
CAS34893-92-0
IUPAC Name1,3-dichloro-5-isocyanatobenzene
Molecular FormulaC7H3Cl2NO
InChI KeyXEFUJGURFLOFAN-UHFFFAOYSA-N
SMILESClC1=CC(=CC(Cl)=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Low melting solid
Infrared spectrumConforms
GC>=95 %
Melting point29°C to 35°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only