Simplify oven loading and unloading with Precision High-Performance Oven Shelves, which are sturdy, steel wire and feature a special non-tipping design.
Have Questions?
Change view
Catalog Number
For Use With (Equipment)
3166179
Precision HP Ovens Models 625/625S
3166180
Precision JP Oven Model 645
3166188
Precision HP Ovens Models 605/605P
3 Options
Catalog number 3166179
Price (INR)
-
For Use With (Equipment):
Precision HP Ovens Models 625/625S
Simplify oven loading and unloading with Thermo Scientific™ Precision™ High-Performance Oven Shelves, which are sturdy, chromium-plated steel wire shelves that feature a special non-tipping design.
Specifications
MaterialSteel
No. of Shelves2 supplied/11 max.
For Use With (Equipment)Precision HP Ovens Models 625/625S