Bis(2-diphenylphosphinoethyl)phenylphosphine, 97%
Bis(2-diphenylphosphinoethyl)phenylphosphine, 97%
Bis(2-diphenylphosphinoethyl)phenylphosphine, 97%
Acros Organics

Bis(2-diphenylphosphinoethyl)phenylphosphine, 97%

CAS: 23582-02-7 | C34H33P3 | 534.56 g/mol
Have Questions?
Catalog NumberQuantityPackaging
316820050
also known as 31682
5 gGlass bottle
Catalog number 316820050
also known as 31682
HSN 29319090
Price (INR)
33,116.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
33,116.00
Each
Chemical Identifiers
CAS23582-02-7
IUPAC Name(2-{[2-(diphenylphosphanyl)ethyl](phenyl)phosphanyl}ethyl)diphenylphosphane
Molecular FormulaC34H33P3
InChI KeyAXVOAMVQOCBPQT-UHFFFAOYSA-N
SMILESC(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or chunks
Elemental Analysis74.1 to 78.7 % (C)
Appearance (Color)White to off-white
Melting point126°C to 132°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only