Tris[2-(diphenylphosphino)ethyl]phosphine, 97%
Tris[2-(diphenylphosphino)ethyl]phosphine, 97%
Tris[2-(diphenylphosphino)ethyl]phosphine, 97%
Acros Organics

Tris[2-(diphenylphosphino)ethyl]phosphine, 97%

CAS: 23582-03-8 | C42H42P4 | 670.69 g/mol
Have Questions?
Catalog NumberQuantity
316830010
also known as 31683
1 g
Catalog number 316830010
also known as 31683
HSN 98020000
Price (INR)
26,649.00
Each
Quantity:
1 g
Request bulk or custom format
Price (INR)
26,649.00
Each
Chemical Identifiers
CAS23582-03-8
IUPAC Name(2-{bis[2-(diphenylphosphanyl)ethyl]phosphanyl}ethyl)diphenylphosphane
Molecular FormulaC42H42P4
InChI KeyTVLNGWSWPKIYAO-UHFFFAOYSA-N
SMILESC(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Melting point134°C to 139°C
Appearance (Form)Powder
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only