Triphenyltin chloride, 95%
Triphenyltin chloride, 95%
Triphenyltin chloride, 95%
Acros Organics

Triphenyltin chloride, 95%

CAS: 639-58-7 | C18H17ClSn | 387.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 317295000
also known as 31729
HSN 29319090
Price (INR)
30,773.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
30,773.00
Each
Chemical Identifiers
CAS639-58-7
IUPAC Nametriphenyltin chloride
Molecular FormulaC18H17ClSn
InChI KeyZDYYCRIXXMWWQC-UHFFFAOYSA-N
SMILESCl.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
Melting point103°C to 109°C
Titration Argentometric94 to 106 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only