Zinc chloride, 99.99%, (trace metal basis), extra pure
Zinc chloride, 99.99%, (trace metal basis), extra pure
Zinc chloride, 99.99%, (trace metal basis), extra pure
Zinc chloride, 99.99%, (trace metal basis), extra pure
Acros Organics

Zinc chloride, 99.99%, (trace metal basis), extra pure

CAS: 7646-85-7 | Cl2Zn | 136.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 318170100
also known as 31817
HSN 28273990
Price (INR)
10,581.00
Each
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
10,581.00
Each
Add to cart
Chemical Identifiers
CAS7646-85-7
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals and/or chunks
Total trace metal impurities=<0.01 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only