MEM α, GlutaMAX™ Supplement, no nucleosides
MEM α, GlutaMAX™ Supplement, no nucleosides
Gibco™

MEM α, GlutaMAX™ Supplement, no nucleosides

Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negativeRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
3256110210 x 500 mL
32561037500 mL
32561029
also known as 32561-029
500 mL
32561094
also known as 32561-094
10 x 500 mL
Catalog number 32561102
Price (INR)
37,900.00
線上優惠
Ends: 31-Mar-2026
56,147.00
Save 18,247.00 (32%)
Each
Add to cart
Quantity:
10 x 500 mL
Customize this product
Price (INR)
37,900.00
線上優惠
Ends: 31-Mar-2026
56,147.00
Save 18,247.00 (32%)
Each
Add to cart

Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells. We offer a variety of Gibco™ MEM α modifications for a range of cell culture applications. Find the right formulation using the media selector tool.


This MEM α is modified as follows:
WithWithout
• Phenol Red• Ribonucleosides
• GlutaMAX™• Deoxyribonucleosides


The complete formulation is available.

Gibco™ MEM α is a modification of Minimum Essential Medium (MEM) that contains non-essential amino acids, sodium pyruvate, lipoic acid, vitamin B12, biotin, and ascorbic acid. MEM α is available without nucleosides for use as a selection medium for DG44 and other DHFR negative cells. Gibco™ MEM α with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. This product is made with Earle’s salts.

MEM α contains no proteins, lipids, or growth factors. Therefore, MEM α requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM α uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineHeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Cell TypePrimary Rat Astrocytes
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco, GlutaMAX
Product TypeMEM α (Minimum Essential Medium α)
Quantity10 x 500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesLow Glucose, Phenol Red, Sodium Pyruvate
Without AdditivesNo HEPES, No Ribonucleosides, No Deoxyribonucleosides
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

Where can I find the osmolality for MEM Medium?

The osmolality is listed in the COA for the particular lot number of the medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

Will depletion, absence, or breakdown of essential growth-promoting components such as glutamine or growth factors reduce the growth rate of my culture?

Yes. If you suspect that this is the case, remove the medium and add fresh medium. Alternatively, you can supplement medium with growth-promoting components. It is also possible to substitute GlutaMax I or II for glutamine in the medium to prevent glutamine exhaustion.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Will the media formulations containing GlutaMAX supplement change with respect to L-glutamine content?

In all media containing GlutaMAX supplement dipeptides as a substitute for L-glutamine, concentration is equimolar with the L-glutamine in the original formulation.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (1)

Citations & References
Abstract
Isolation, characterization and propagation of mitotically active germ cells from adult mouse and human ovaries.
Authors:Woods DC, Tilly JL
Journal:Nat Protoc
PubMed ID:23598447
Accruing evidence indicates that production of new oocytes (oogenesis) and their enclosure by somatic cells (folliculogenesis) are processes not limited to the perinatal period in mammals. Endpoints ranging from oocyte counts to genetic lineage tracing and transplantation experiments support a paradigm shift in reproductive biology involving active renewal of oocyte-containing ... More