n-Methyloctylamine, 98%
n-Methyloctylamine, 98%
n-Methyloctylamine, 98%
Acros Organics

n-Methyloctylamine, 98%

CAS: 2439-54-5 | C9H21N | 143.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 325620250
also known as 32562
HSN 29211990
Price (INR)
26,394.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
26,394.00
Each
Chemical Identifiers
CAS2439-54-5
IUPAC Namemethyl(octyl)amine
Molecular FormulaC9H21N
InChI KeySEGJNMCIMOLEDM-UHFFFAOYSA-N
SMILESCCCCCCCCNC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Titration with HClO4>=97.5 %
Refractive index1.4260 to 1.4290 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only