n-Hexane, for pesticide residue analysis, approx. 95% n-Hexane
n-Hexane, for pesticide residue analysis, approx. 95% n-Hexane
n-Hexane, for pesticide residue analysis, approx. 95% n-Hexane
Acros Organics

n-Hexane, for pesticide residue analysis, approx. 95% n-Hexane

CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number 326660025
HSN 29011000
Price (INR)
10,273.00
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS110-54-3
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Residue after evaporation=<2 ppm
GC(ECD) Passes test
GC(FID) Passes test
GC(NPD) Passes test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
RUO – Research Use Only