Acetonitrile, 99.9+%, Extra Dry, AcroSeal™
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™
Acros Organics

Acetonitrile, 99.9+%, Extra Dry, AcroSeal™

Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methylkyanid, methane, cyano, methyl cyanide, mecn, methanecarbonitrile, ethanenitrile, ethyl nitrile, cyanure de methyl, cyanomethane, acetonitrile, 100mL, 0.002 % max., Vapor Pressure: 97mbar at 20 deg.C, 99.9+%, 02, 183, 81.0 deg.C to 82.0 deg.C | CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Catalog number 326811000
HSN 29269000
Price (INR)
11,112.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (INR)
11,112.00
Each
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Residue after evaporation=<0.0005 %
Infrared spectrumConforms
Water=<0.001 % (Coulometric)
Acetamide=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only