1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™
Acros Organics

1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™

CAS: 872-50-4 | C5H9NO | 99.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 326931000
HSN 29337900
Price (INR)
6,606.00
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (INR)
6,606.00
Each
Chemical Identifiers
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<20 APHA
GC>=99.4 %
Water=<0.005 %
Infrared spectrumConforms

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only