N,N-Dimethylformamide, 99.8%, for molecular biology, DNAse, RNAse and Protease free
N,N-Dimethylformamide, 99.8%, for molecular biology, DNAse, RNAse and Protease free
N,N-Dimethylformamide, 99.8%, for molecular biology, DNAse, RNAse and Protease free
Acros Organics

N,N-Dimethylformamide, 99.8%, for molecular biology, DNAse, RNAse and Protease free

CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
10 L
Catalog number 327171000
HSN 29241900
Price (INR)
5,871.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (INR)
5,871.00
Each
Chemical Identifiers
CAS68-12-2
IUPAC NameN,N-dimethylformamide
Molecular FormulaC3H7NO
InChI KeyZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILESCN(C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.8 %
Water=<0.05 % (Coulometric)
UV(undiluted)
UVat 270 nm A: =<1
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For molecular biology
RUO – Research Use Only