Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3
Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3
Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3
Acros Organics

Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3

CAS: 610769-35-2 | C14H30BN3O14 | 475.211 g/mol
Have Questions?
Catalog NumberQuantity
327330010
also known as 32733
1 L
Catalog number 327330010
also known as 32733
HSN 29221990
Price (INR)
6,936.00
Each
Quantity:
1 L
Request bulk or custom format
Price (INR)
6,936.00
Each
Chemical Identifiers
CAS610769-35-2
IUPAC Name2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid; 2-amino-2-(hydroxymethyl)propane-1,3-diol; boric acid
Molecular FormulaC14H30BN3O14
InChI KeyOSBLTNPMIGYQGY-UHFFFAOYSA-N
SMILESOB(O)O.NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
pH8.2 to 8.4 (at 25°C)
Boric acid0.089 M
Dnase,Rnase,Protease activityNone detected
Tris-Base0.089 M
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent, pH buffer
RUO – Research Use Only

Frequently asked questions (FAQs)

My Thermo Scientific brand TBE buffer has precipitated out of solution. What can I do?

Concentrations of 5X Tris Borate EDTA (TBE) buffer or greater may precipitate due to the nature of the product. This precipitation is a result of nucleation of salt crystals by dust particles or other insoluble materials. In some cases, we recommend heating the buffer to ~ 50°C to re-dissolve the TBE into solution. Alternately, to avoid precipitation of high concentration TBE buffers, after the solution is prepared, you can filter it through a 0.2 µm cellulose acetate or cellulose nitrate filter. These treatments have no deleterious effect on the buffering properties of TBE.