Genistein, 99%, synthetic
Genistein, 99%, synthetic
Genistein, 99%, synthetic
Acros Organics

Genistein, 99%, synthetic

CAS: 446-72-0 | C15H10O5 | 270.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
25 mg
Packaging:
Glass bottle
Catalog number 328271000
HSN 29329900
Price (INR)
35,555.00
Each
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
35,555.00
Each
Chemical Identifiers
CAS446-72-0
IUPAC Name5,7-dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
Molecular FormulaC15H10O5
InChI KeyTZBJGXHYKVUXJN-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Color)White to light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only