Glycerol, 99+%, pure, synthetic
Glycerol, 99+%, pure, synthetic
Glycerol, 99+%, pure, synthetic
Acros Organics

Glycerol, 99+%, pure, synthetic

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 332031000
HSN 29054500
Price (INR)
4,782.00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS56-81-5
IUPAC Namepropane-1,2,3-triol
Molecular FormulaC3H8O3
InChI KeyPEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILESOCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Clear viscous liquid
Color scale=<10 APHA
GC>=99.0 %
Refractive index1.4700 to 1.4750 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only