Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.9)
Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.9)
Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.9)
Acros Organics

Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.9)

CAS: 9004-32-4 | (C12 H14 O9 R6)n | 263.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
Packaging:
View all
Plastic bottle
Catalog number 332620010
HSN 39123100
Price (INR)
22,559.00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS9004-32-4
IUPAC Name2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
Molecular Formula(C12 H14 O9 R6)n
InChI KeyDPXJVFZANSGRMM-UHFFFAOYNA-N
SMILES[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Lead (Pb)=<3 ppm
Appearance (Form)Crystalline powder
Loss on drying=<10 % (As packed) (3 to 5 g, 105°C, 2 h)
Viscosity1500 to 3100 mPa.s (2 % at 25°C) (Brookfield)
Cadmium (Cd)=<1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only