(1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine, 95%
(1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine, 95%
(1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine, 95%
Acros Organics

(1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine, 95%

CAS: 68737-65-5 | C8H20N2 | 144.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 337830010
also known as 33783
HSN 29213090
Price (INR)
44,976.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
44,976.00
Each
Chemical Identifiers
CAS68737-65-5
IUPAC Name(1R,2R)-N1,N2-dimethylcyclohexane-1,2-bis(aminium)
Molecular FormulaC8H20N2
InChI KeyJRHPOFJADXHYBR-HTQZYQBOSA-P
SMILESC[NH2+][C@@H]1CCCC[C@H]1[NH2+]C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Low melting solid
Infrared spectrumConforms
GC>=94 %
Specific optical rotation-137° to -149° (20°C, 589 nm) (C=1, CHCl3)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only