Product Image
Thermo Scientific™
Dynabeads™ M-270 Epoxy, for OEM and industrial use only
Dynabeads™ M-270 Epoxy are monosized 2.8-μm superparamagnetic polystyrene beads coated with a cross-linked layer of hydrophilic polyether.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
34004D10 g
34003D1 g
Catalog number 34004D
Price (INR)
-
Quantity:
10 g
Dynabeads™ M-270 Epoxy are monosized 2.8-μm superparamagnetic polystyrene beads coated with a cross-linked layer of hydrophilic polyether. Epoxy functional groups allow for covalent coupling of antibodies or other proteins, peptides, lectins, haptens, or functional enzymes onto a non-charged bead surface through amine or thiol groups with no further activation of the surface. No blocking protein is needed during the preparation, and standard assay diluents may be used. The Dynabeads™-ligand complex can be used for immunoassays or any other application requiring an active biomolecule conjugated to a magnetic solid support.
Dynabeads™ magnetic beads are widely used as a solid-phase for presenting antibodies or antigens in automated clinical immunoassay systems, where the combination of specific antigens or antibodies and the superparamagnetic properties of the beads provide rapid reaction kinetics both in the coating process, separation, and during washing of the analyte.
The product is provided in a freeze-dried format and meets reputable Dynal high standards with respect to reproducibility and automation ability, and drives reliability for your assays.
Benefits:
  • Efficient and reproducible immobilization of antibodies (or other ligands)
  • Easy and reproducible handling in manufacturing
  • Rapid separation and washing
  • Reproducible behavior in automation
  • Easy coupling without need for protein blocking
  • Freeze-dried beads
  • High activity of bound antibody and antigen
  • Easy assay optimization
  • Well-suited for binding of small antigens such as synthetic or recombinant peptides
  • Generally low background and high signal-to-noise ratio
Applications:
Ideally suited for magnetic bead-based immunoassays requiring a low background signal. Also suited for any application involving immunoaffinity binding to antibody or antigen immobilized on a magnetic surface.
Note:
This particular product format is for large volume customers, available on an OEM basis. This product is also available in smaller volumes (14301D and 14302D).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineDynabeads
Quantity10 g
Surface FunctionalityEpoxy, Hydrophilic
Diameter (Metric)2.8 μm
MaterialPolystyrene
Product TypeEpoxy Bead
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).
Frequently asked questions (FAQs)
What reactive groups do you offer for your surface-activated Dynabeads magnetic beads?
We offer tosyl, epoxy, carboxylic acid and amine activated Dynabeads magnetic beads. Please see the link (https://tools.thermofisher.com/content/sfs/brochures/Surface_Activated_Dynabeads.PDF) for a comparison of the beads.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
I am using surface-activated Dynabeads magnetic beads. When do I need to use a linker secondary antibody?
A linker antibody provides proper orientation of the target-specific primary antibody. Optimal orientation of the primary antibody is more important for reacting with larger organelles than for small organelles or membrane fractions. Different linkers can be used, but we recommend using an Fc-binding antibody, such as a monoclonal or polyclonal anti-mouse IgG. The linker antibody must be affinity purified and not contain stabilizers such as sugars or proteins that may bind to the Dynabeads magnetic beads. The specific primary antibody, if polyclonal, must be affinity purified in order to provide a high density of the specific antibody on the Dynabeads magentic beads surface.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Which surface-activated Dynabeads magnetic beads should I choose for immobilizing my protein?
This depends on the nature of the specific ligand to be immobilized and the desired downstream application.
-The most frequently used surface-activated Dynabeads magnetic beads for protein isolation are the Tosyl-activated Dynabeads magnetic beads M-280. These beads are hydrophobic, easy to handle and ideal for covalent binding of antibodies for immunoprecipitation. Other ligands could also be covalently bound to these beads, but they have to tolerate conditions like neutral to high pH and high temperatures (required for covalent bond formation).
-Dynabeads magnetic beads M-270 Epoxy are used when the ligand to be immobilized needs to be treated gently and will not tolerate harsh binding conditions like high temperature or pH. Proteins/peptides (other than antibodies) and enzymes are often coupled onto these beads.
-Dynabeads magnetic beads M-270 Amine is often used in combination with crosslinkers to create specific surface groups on the beads. Hetro-bifunctional crosslinkers with an amine-reactive NHS group at one end and another chemical group of your choice at the other end are most frequently used. For example, Dynabeads magnetic beads M-270 Amine can be reacted with a hetero-bifunctional crosslinker containing a NHS group at one end and maleimide at the other end to create Dynabeads magnetic beads with a maleimide surface. Since maleimide reacts specifically with sulfhydryl groups, these modified Dynabeads magnetic beads can be used for applications where binding of sulfhydryl groups are desired. Dynabeads magnetic beads M-270 Amine may also be used for direct ligand coupling via aldehyde or ketone groups by Schiff base (imine) formation and reductive amination. In addition carboxylic acid groups on a ligand can be activated with a carbodiimide like EDC, which results in a direct amide bond formation between the beads and the ligand. Alternatively, a crosslinker may be introduced to the ligand. After activation of the ligand with crosslinker, the free end on the crosslinker has to be amine reactive.
-Dynabeads magnetic beads M-270 Carboxylic Acid and Dynabeads magnetic beads MyOne Carboxylic Acid can also be used for immobilizing proteins. The carboxylic acid groups on these beads need to be activated with a carbodiimide before coupling. The negatively-charged surface of these beads may attract positively charged proteins and cause nonspecific binding. This needs to be considered if these beads are going to be used for immunoprecipitation.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
How are proteins coupled to surface-activated Dynabeads magnetic beads?
Dynabeads M-280 Tosyl activated magnetic beads allow direct covalent binding to primary amino or sulfhydryl groups in proteins and peptides at high pH and high temperature.
-Dynabeads M-270 Epoxy magnetic beads allow direct covalent binding to primary amino and sulfhydryl groups in proteins and peptides at neutral pH and across a wide temperature range.
-Dynabeads M-270 Amine magnetic beads allow direct covalent binding through reductive amination of aldehydes, or the use of bifunctional amine-reactive crosslinkers.
-Dynabeads M-270 and MyOne Carboxylic Acid magnetic beads allow covalent amide formation with primary amino groups in proteins and peptides.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What do the designations M-280, M-270, and MyOne mean on Dynabeads magnetic beads?
The M stands for magnetic. M-280 refers to hydrophobic 2.8 micron beads, while M-270 refers to hydrophilic 2.8 micron beads. MyOne refers to 1 micron beads.
Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.