1-Step™ TMB ELISA Substrate Solutions
Thermo Scientific™

1-Step™ TMB ELISA Substrate Solutions

Thermo Scientific™ Pierce TMB ELISA Substrates detect horseradish peroxidase (HRP) activity, yielding a blue color (Amax = 370 nm and 652 nm) that changes to yellow (Amax = 450 nm) upon addition of a sulfuric or phosphoric acid stop solution.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitySubstrate
340291 LUltra TMB-ELISA Substrate
34024250 mLSlow TMB-ELISA Substrate
34022250 mLTurbo TMB-ELISA Substrate
34028250 mLUltra TMB-ELISA Substrate
34021400 mL KitTMB Substrate
Catalog number 34029
Price (INR)
55,041.00
Online offer
Ends: 31-Dec-2026
78,629.00
Save 23,588.00 (30%)
Each
Quantity:
1 L
Substrate:
Ultra TMB-ELISA Substrate
Request bulk or custom format
Price (INR)
55,041.00
Online offer
Ends: 31-Dec-2026
78,629.00
Save 23,588.00 (30%)
Each

TMB (3,3',5,5' tetramethylbenzidine) is a chromagen that yields a blue color when oxidized with hydrogen peroxide (catalyzed by HRP) with major absorbances at 370 nm and 652 nm. The color then changes to yellow (measured at 450 nm) upon addition of sulfuric or phosphoric acid to stop the reaction. TMB is very sensitive and more quickly oxidized than other HRP substrates, resulting in faster color development. Unlike other commercially available substrates, these products contain no DMF or DMSO.

There are three formulations that differ primarily in their sensitivities. Slow TMB is intermediate in sensitivity, ideal for kinetic readings. The sensitivity of the Turbo TMB compares to that of OPD used at approximately 1 mg/mL. Ultra TMB-ELISA produces the highest signal-to-noise ratio and sensitivity in the picogram range.

Thermo Scientific Pierce 1-Step TMB ELISA Substrates detects horseradish peroxidase (HRP) activity, yielding a blue color (Amax = 370 nm and 652 nm) that changes to yellow (Amax = 450 nm) upon addition of a sulfuric or phosphoric acid stop solution.

Features of 1-Step TMB ELISA Substrates:
• Available in traditional or ready-to-use formulations
• No additional reagents or filtering required
• Non carcinogenic
• Various levels of sensitivity and development speed to suit any assay
• No DMF or DMSO present in the reagents

TMB is the most popular chromogenic substrate for HRP detection in ELISA and is available in several formats. The Pierce 1-Step TMB Substrates are one-component substrates that require no preparation before use. Unlike other commercially available substrates, these products contain no DMF or DMSO.

Thermo Scientific Pierce 1-Step Ultra TMB yields the greatest sensitivity among the TMB substrates, followed by 1-Step Turbo TMB and 1-Step Slow TMB. The sensitivity of the 1-Step Turbo TMB is similar to OPD-based substrates formulated at approximately 1 mg/mL. 1-Step Slow TMB has intermediate sensitivity—more sensitive than ABTS, but less sensitive than o-phenylenediamine dihydrochloride (OPD) or 1-Step Turbo TMB. The 1-Step Slow TMB is an ideal substrate for kinetic studies.

TMB (3,3',5,5'-tetramethylbenzidine) is a chromogen that yields a blue color when oxidized, typically as a result of oxygen radicals produced by the hydrolysis of hydrogen peroxide by HRP. For kinetic or non-stopped ELISA assays, the TMB chromogen has maximal absorbances at 370 nm and 652 nm. The color then changes to yellow with the addition of sulfuric or phosphoric acid with maximum absorbance at 450 nm. A green reaction product may result from partial conversion to the yellow product from the blue intermediate. TMB is very sensitive ELISA substrate and is more quickly oxidized than other HRP substrates, resulting in faster color development.

For Research Use Only. Not for use in diagnostic procedures.

1-Step Substrates are available in three formulations that vary in sensitivity:

  • 1-Step Slow TMB has intermediate sensitivity – more sensitive than ABTS, but less sensitive than OPD or 1-Step Turbo TMB
  • 1-Step Turbo TMB has sensitivity comparable to OPD used at approximately 1mg/mL
  • 1-Step Ultra TMB-ELISA has the highest signal-to-noise ratio and sensitivity in the picogram range

General References:

  1. Josephy, P.D., et al. (1982). J. Biol. Chem. 257(7), 3669-3675.
  2. Josephy, P.D., et al. (1983). J. Biol. Chem. 258(9), 5561-5569.
  3. Liem, H.H., et al. (1979). Anal. Biochem. 98, 388-393.
Specifications
Detection MethodColorimetric
Product Line1-Step
Shipping ConditionAmbient
SubstrateUltra TMB-ELISA Substrate
Substrate PropertiesChemical Substrate
Substrate TypeHRP (Horseradish Peroxidase)
Product TypeELISA Substrate
Quantity1 L
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C, protected from moisture.

Frequently asked questions (FAQs)

Are all your TMB substrates transparent and do they all turn blue after reacting with the HRP enzyme of an antibody?

No, we offer different TMB substrates for different applications.
We recommend the following TMB substrates for this particular application:
- eBioscience TMB Solution (1X) (Cat. No. 00-4201-56)
- (For higher sensitivity) 1-Step Ultra TMB-ELISA Substrate Solution (Cat. No. 34029)

What is the shelf-life of 1-Step Ultra TMB-ELISA Substrate Solution?

The 1-Step Ultra TMB-ELISA Substrate Solution is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

My lab doesn't have a fluorescent or chemiluminescent plate reader and I need the most sensitive substrate for Horseradish Peroxidase (HRP). Which of your colorimetric substrates should I use?

The 1-Step Ultra TMB-ELISA is our most sensitive substrate for HRP in plate-based assays. It can detect down to ~ 2 pg/well or 20 pg/mL.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can the 1-Step Ultra TMB-ELISA Substrate Solution be used as a precipitating chromogen for Western Blot, Immunohistochemistry and immunocytochemistry?

No. This product is formulated to generate a soluble, colored product that absorbs light at 450 nm. When used properly, there will be no precipitate, therefore, this substrate is only appropriate for plate-based assays, such as ELISA.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Is the 1-Step Ultra TMB-ELISA Substrate Solution an appropriate substrate for kinetic assays?

No. Although the non-stopped substrate will turn a blue color in the presence of Horseradish Peroxidase and absorb light at 307 nm and 652 nm, color development proceeds very quickly. In the time that it takes to read the full plate, the absorbance of the first wells will have changed. It is better to stop the the color development in all wells at approximately the same time point with an acidic soluton, turning it yellow and then read the absorbance at 450 nm.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.