Low DNA Binding Snap Cap Microcentrifuge Tube
Thermo Scientific™
Low DNA Binding Snap Cap Microcentrifuge Tube
Green features
Microcentrifuge Tubes ensure optimal sample integrity with minimal DNA loss. These tubes provide secure closure and reliable performance, perfect for sensitive applications in molecular biology.
Have Questions?
Change viewbuttonViewtableView
Volume (Metric):
0.6 mL
1.5 mL
2 mL
Catalog number 3404-DLBPK
Price (INR)
-
Volume (Metric):
1.5 mL
Request bulk or custom format
Specifications
Green FeaturesACT Ecolabel (Silver)
Product TypeMicrocentrifuge Tube
ShapeCylindrical
Volume (Metric)1.5 mL
AutoclavableAutoclavable
View more
The importance of low DNA binding microcentrifuge tubes cannot be overstated. These tubes are specifically engineered to minimize the adhesion of DNA to the tube walls, which is crucial in preserving the quantity and quality of DNA samples during various laboratory procedures.
Ensuring that maximum DNA recovery is achieved and reducing the risk of sample loss and contamination. Particularly vital in applications such as PCR, sequencing, and cloning, where even small losses can significantly impact the results. Additionally, the assurance that these tubes are free from PCR inhibitors, UV leachables, and human DNA further enhances the reliability of experimental outcomes by preventing any potential interference with sensitive assays.