IPTG
Thermo Scientific™

IPTG

Thermo Scientific Pierce IPTG is isopropyl-β-D-thiogalactopyranoside for use with beta-galactosidase substrates such as X-Gal to detect expression of vectors, plasmids,Read more
Have Questions?
Catalog NumberQuantity
340601 g
Catalog number 34060
Price (INR)
9,702.00
Online offer
Ends: 31-Dec-2026
14,372.00
Save 4,670.00 (32%)
Each
Quantity:
1 g
Request bulk or custom format
Price (INR)
9,702.00
Online offer
Ends: 31-Dec-2026
14,372.00
Save 4,670.00 (32%)
Each
Thermo Scientific Pierce IPTG is isopropyl-β-D-thiogalactopyranoside for use with beta-galactosidase substrates such as X-Gal to detect expression of vectors, plasmids, or DNA fragments that contain the reporter gene.

The reagent X-Gal (5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside) is a chromogenic substrate for β-galactosidase that yields a blue precipitate, is soluble in DMF, and can be used to detect vectors, plasmids, or DNA fragments that contain the β-Galactosidase gene.

IPTG (Isopropyl-β-D-thiogalactopyranoside) may be used to maximize the expression of genes in expression vectors. When screening a lambda bacteriophage library with an antibody, the probability of success can be increased by preventing the expression of the fusion protein until the plaques are well established. By placing a nitrocellulose filter containing the inducer IPTG onto the plate and continuing growth at 37°C, the expression of the potential β-galactosidase-cDNA fusion protein can be induced after 3-4 hours of plaque growth. The nitrocellulose filter is then screened with antibodies. IPTG is also used in combination with X-Gal to detect active β-galactosidase and is practical in applications using bacteriophage M13 and Igt11 vectors.

Features of IPTG:

• X-Gal yields an intense blue precipitate to detect β-galactosidase expression
• Can be combined with IPTG to selectively maximize the detection of β-galactosidase
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeIPTG (Isopropyl β-D-thiogalactopyranoside)
Quantity1 g
FormPowder
Unit SizeEach
Contents & Storage
Store at or below -20°C.