PCR Plate Strip Caps
PCR Plate Strip Caps
PCR Plate Strip Caps
Thermo Scientific™

PCR Plate Strip Caps

Ensure secure seals with these cap strips, which fit 24-, 48- and 96-well plates and 0.2mL tube strips
Have Questions?
Catalog NumberColor
3418FNatural
Catalog number 3418F
Price (INR)
-
Color:
Natural
Ensure secure seals by using Thermo Scientific™ PCR Plate Strip Caps, which fit 24-, 48- and 96-well plates and 0.2mL tube strips
  • Certified DNA-, DNase, RNase and pyrogen-free
  • Natural color
Specifications
For Use With (Application)Provide secure seals
Product TypePCR Plate Strip Cap
ColorNatural
No. of Wells12
Plate Compatibility24/48/96-well Plate, 0.2 mL Tube Strips
Unit SizeEach