Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, 98%, Stabilized
Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, 98%, Stabilized
Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, 98%, Stabilized
Acros Organics

Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, 98%, Stabilized

CAS: 1025-15-6 | C12H15N3O3 | 249.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 342125000
also known as 34212
HSN 29336990
Price (INR)
22,370.00
Each
Quantity:
500 g
Request bulk or custom format
Price (INR)
22,370.00
Each
Chemical Identifiers
CAS1025-15-6
IUPAC Nametris(prop-2-en-1-yl)-1,3,5-triazinane-2,4,6-trione
Molecular FormulaC12H15N3O3
InChI KeyKOMNUTZXSVSERR-UHFFFAOYSA-N
SMILESC=CCN1C(=O)N(CC=C)C(=O)N(CC=C)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5120 to 1.5140 (20°C, 589 nm)
Stabilizer100 to 1200 ppm MEHQ
Appearance (Color)Clear colorless to yellow
Appearance (Form)Viscous liquid or solid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only