6-Hydroxycaproic acid, 95%
6-Hydroxycaproic acid, 95%
6-Hydroxycaproic acid, 95%
Acros Organics

6-Hydroxycaproic acid, 95%

CAS: 1191-25-9 | C6H12O3 | 132.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
342240010
also known as 34224
1 gGlass bottle
Catalog number 342240010
also known as 34224
HSN 29181990
Price (INR)
15,555.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
15,555.00
Each
Chemical Identifiers
CAS1191-25-9
IUPAC Name6-hydroxyhexanoate
Molecular FormulaC6H11O3
InChI KeyIWHLYPDWHHPVAA-UHFFFAOYSA-M
SMILESOCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Additional infoMay contain varying amounts of dimers
Additional infoMay contain varying amounts of water
Additional infoMay contain varying amounts of dimers
Additional infoMay contain varying amounts of water
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only