Microtiter Tube Plug Strip
Microtiter Tube Plug Strip
Actual product may vary
Microtiter Tube Plug Strip
Seal 1.2mL microtiter tubes tightly for storage using these polyethylene plug strips.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityQuantity
3425-11Sterile120/Pk., 5/Cs.
3426-11Non-sterile120/Pk., 5/Cs.
Catalog number 3425-11
Price (INR)
11,941.00
Each
Add to cart
Sterility:
Sterile
Quantity:
120/Pk., 5/Cs.
Price (INR)
11,941.00
Each
Add to cart
Seal 1.2mL microtiter tubes tightly for storage by using Thermo Scientific™ Microtiter Tube Plug Strips.
  • LDPE non-autoclavable
  • Eight-plug strips can be easily cut apart for individual samples
  • Available sterile and nonsterile
Specifications
Product Type8-cap Strip
AutoclavableNot Autoclavable
Description1.2mL Microtiter Tube 8-cap plug strip
MaterialLDPE
ColorNatural
Packaging10 sealed bags of twelve 8 cap strips/unit, 5 units/case
Quantity120/Pk., 5/Cs.
SterilitySterile
For Use With (Equipment)Plug Strips
Unit SizeEach