Nalgene™ PPCO High Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack
Nalgene™ PPCO High Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack
Actual product may vary
Nalgene™ PPCO High Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack
Nalgene™ PPCO High Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack
Nalgene™ PPCO High Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack
Nalgene™ PPCO High Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack
Reduce the risk of contamination with high-profile PPCO closures with color coders that are free of toxic heavy metals and have no O-rings that can accidentally fall out.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
342820-0111White
342820-0110Clear (no coder)
342820-0112Yellow
342820-0113Orange
342820-0114Green
342820-0115Red
342820-0116Blue
342820-0117Gold
342820-0118Purple
342820-0119Natural
342820-1110Teal
Catalog number 342820-0111
HSN 39235090
Price (INR)
27,672.00
Case
Add to cart
Color:
White
Price (INR)
27,672.00
Case
Add to cart
Reduce the risk of contaminating contents using Thermo ScientificTM NalgeneTM PPCO High Profile Closures with Color Coders for Micro Packaging Vials. Polypropylene copolymer closures have no O-ring to fall out and contaminate content, and they are offered in a variety of colors.
Features:
  • Vials (listed separately) and closures are pressure-tested together at 7.5 psig.
  • Permanently attached, color-coded inserts are free of toxic heavy metals and do not come in contact with vial contents.
  • Threaded, screw closures are sterile to 10-6 SAL and RNase/DNase free
  • Leakproof† when used with Nalgene Micro Packaging Vials.
Ordering Information:
Micro Packaging Vial Closures are bulk packed without vials, 500 per inner bag, 1,000 per case. Order sterile vials separately (Cat. Nos. 342800, 342805).
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database
Specifications
DescriptionHigh-profile closures w/color coders for micro packaging vials
ColorWhite
Diameter (Metric) Outer11 mm
MaterialPPCO
SterilitySterile
No. per Case1000
No. per Pack500
Product LineNalgene
Closure MaterialPPCO
Closure TypeScrew Cap
Quantity500 Bag, 1000 Cs.
Closure Size11 mm
Certifications/ComplianceISO 13485:2016
Product TypeClosure
Unit SizeCase
Frequently asked questions (FAQs)
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.