Nalgene™ Coex Polyethylene Film Tank Liners, 80 gallon
Nalgene™ Coex Polyethylene Film Tank Liners, 80 gallon
Thermo Scientific™
Nalgene™ Coex Polyethylene Film Tank Liners, 80 gallon
Enhance mixing capabilities open-bag, flat-bottom Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners , which offer cost-effective, single-use fluid processing.
Have Questions?
Catalog number 343050-0080
Price (INR)
-
Request A Quote
Enhance mixing capabilities with open-bag, flat-bottom liners. Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene cylindrical plastic tanks. Coex Polyethylene film is free of animal-derived components (ADCF).
  • Designed to fit Nalgene cylindrical tanks from 5 gal (19L) to 200 gal (787L)
  • Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic
Specifications
Capacity (English)80 gal.
For Use With (Equipment)Nalgene cylindrical tanks from 5 gal (19L) to 200
No. per Case10
No. per Pack10
Capacity (Metric)303 L
MaterialPolyethylene
TypeTank Liner
Unit SizeCase of 10