Nalgene™ Coex Polyethylene Film Tank Liners
Thermo Scientific™

Nalgene™ Coex Polyethylene Film Tank Liners

Enhance mixing capabilities open-bag, flat-bottom Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners , which offer cost-effective, single-use fluid processing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)
343050-0200200 gal.
343050-00055 gal.
343050-00077.5 gal.
343050-001010 gal.
343050-003030 gal.
343050-005555 gal.
343050-0100100 gal.
343050-0150150 gal.
Catalog number 343050-0200
Price (INR)
-
Capacity (English):
200 gal.

Enhance mixing capabilities with open-bag, flat-bottom liners. Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene cylindrical plastic tanks.

  • Designed to fit Nalgene cylindrical tanks from 5 gal (19L) to 200 gal (787L)
  • Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic

Warranty:
90 days

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications
Capacity (English)200 gal.
For Use With (Equipment)Nalgene cylindrical tanks from 5 gal (19L) to 200
No. per Case10
No. per Pack10
Capacity (Metric)757 L
MaterialPolyethylene
TypeTank Liner
Unit SizeCase of 10