1-Butanesulfonic acid, sodium salt, 99+%, HPLC grade
1-Butanesulfonic acid, sodium salt, 99+%, HPLC grade
1-Butanesulfonic acid, sodium salt, 99+%, HPLC grade
Acros Organics

1-Butanesulfonic acid, sodium salt, 99+%, HPLC grade

CAS: 2386-54-1 | C4H9NaO3S | 160.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 343800050
also known as 34380
HSN 29041090
Price (INR)
6,249.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
6,249.00
Each
Chemical Identifiers
CAS2386-54-1
IUPAC Namesodium butane-1-sulfonate
Molecular FormulaC4H9NaO3S
InChI KeyXQCHMGAOAWZUPI-UHFFFAOYSA-M
SMILES[Na+].CCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
UV(5.5 % in water, pH 3) 1 cm cell
UVat 220 nm A: =<0.1
UVat 280 nm A: =<0.04
Appearance (Color)White
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ion pair chromatography
RUO – Research Use Only