4-Hydroxypyridine, 95%
4-Hydroxypyridine, 95%
4-Hydroxypyridine, 95%
Acros Organics

4-Hydroxypyridine, 95%

CAS: 626-64-2 | C5H5NO | 95.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 348835000
also known as 34883
HSN 29333919
Price (INR)
82,803.00
Each
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
82,803.00
Each
Chemical Identifiers
CAS626-64-2
IUPAC Name1,4-dihydropyridin-4-one
Molecular FormulaC5H5NO
InChI KeyGCNTZFIIOFTKIY-UHFFFAOYSA-N
SMILESO=C1C=CNC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=94 %
HPLC>=94 %
Melting point139°C to 151°C
Appearance (Color)White to yellow to beige to light brown
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only