Screw Cap Micro Tubes
Thermo Scientific™

Screw Cap Micro Tubes

Choose these tubes, which available in conical and free-standing designs when seeking secure, short-term sample storage in low temperatures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Bottom ShapeClosure TypePackaging
3488PK
also known as 3488
2 mLSkirtedScrew Cap with O-ring (Unattached)500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
3422-11PK
also known as 3422-11
0.5 mLSkirtedNot Included500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
3465PK0.5 mLSkirtedAttached Cap500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
34781.5 mLSkirtedNot Included500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
3464PK
also known as 3464
1.5 mLNon-skirtedScrew Cap with O-ring (Unattached)500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
34611.5 mLNon-skirtedAttached Cap500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
34902 mLSkirtedNot Included500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
3468PK
also known as 3468
2 mLNon-skirtedScrew Cap with O-ring (Unattached)500/Pack
34632 mLNon-skirtedAttached Cap500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
Catalog number 3488PK
also known as 3488
Price (INR)
4,902.00
Pack of 500
Capacity (Metric):
2 mL
Bottom Shape:
Skirted
Closure Type:
Screw Cap with O-ring (Unattached)
Packaging:
500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
Price (INR)
4,902.00
Pack of 500

Choose Thermo Scientific™ Screw Cap Micro Tubes, which available in conical and free-standing designs, when seeking secure, short-term sample storage in low temperatures. Their robust design and material selection are well suited for work with biologics and offer rich features, including ultra-clear, externally threaded tubes and UV protective amber tubes. Options include: with or without a self-standing skirted bottom, with or without tethered straps, non-knurl and knurled tubes for single-hand operation, choice of multicolored caps and volume sizes of 0.5mL, 1.5mL and 2.0mL sizes. Screw Cap Micro Tube Caps offer a durable design incorporating an O-ring that helps prevent leaks under demanding conditions.

Non-Sterile is free of Rnase/Dnase, Human and Mouse DNA, Pyrogen & Endotoxin, ATP, PCR Inhibitor.
Sterile E-Beam is free of Rnase/Dnase, Human and Mouse DNA, Pyrogen, Endotoxin, ATP and PCR Inhibitor.
Packaging is tailored to meet broad requirements. Tubes are packaged with and without caps, caps are attached or unattached, placed in a single bag and double bagged feature for clean room environments is available upon request.
100% virgin polypropylene resin specifically selected for high clarity and biologic applications.
O-ring design, thickness and material selection have been optimized to provide protective features for maximum sealing ability, biologic interaction and temperature capability. The black colored O-ring is made from EPDM.
Automated System Compatible. Whether automate is used for filling, capping or labeling, the product is designed to withstand high torque pressures associated with automation. Product design helps to address common automated issues like: fissure cracks, bowl feeder jam, over/under torqued caps and cross threading.
Volume range of 0.5, 1.5 and 2.0mL.
Tube Color choices are an ultra-high clarity, natural color or amber option which provides ultra violet (UV) protection of 1.22% transmittance between 220 and 500nm.
Standard or Tethered cap, all available in a rainbow of colors. Visit cap product page for additional information.
Knurl tubes feature the ability to lock tube into specialty racks for single-hand operation.
Non knurl tubes ideal for automation and labeling, tubes are plain, removed of any labeling interference.
Skirted or Self Standing tubes can stand upright on their own, no rack required.
Conical tubes requires a rack to support them in the upright position.
Low nucleic acid and protein binding and low enzyme inhibition are features that Resin selection features.
RCF range from 18,000 to 25,000 x g.
Temperature Range -80°C to +95°C. Short-term storage in -80°C, -20°C, dry ice and wet ice. Short-term storage in vapor phase LN2. Suitable for a quick dip flash freeze in LN2.
Water Bath compatible up to +95°C.

† Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.

Screw cap micro tubes and caps are not recommended for autoclave. Sterile options available.

Not intended to submerge and store in liquid phase LN2.

Specifications
Binding PropertyLow Nucleic Acid and Protein Binding & Low Enzyme Inhibition
Clean ClaimsNon-Sterile Free of: Rnase/Dnase, Human and Mouse DNA, Pyrogen & Endotoxin, ATP, PCR Inhibitor
IncludesKnurl
Max. TorqueManual/Hand Torque: Standard: 0.125 lbsf.in to 2.5 lbsf.in; Metric: 1.412 Ncm to 28.24 Ncm. Automated/Robotic Torque Standard: 1.312 lbsf.in to 2.5 lbsf.in; Metric: 14.83 Ncm to 28.24 Ncm
Packaging500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
Product TypeScrew-cap Microtube
Temperature Range (Metric)Cold Storage: -80° to +95°C. Liquid Nitrogen: Short term Vapor Phase & Quick Flash Freeze acceptable, not recommmended for LN2 submersion storage
AutoclavableNot Autoclavable
Bottom ShapeSkirted
Capacity (Metric)2 mL
Closure TypeScrew Cap with O-ring (Unattached)
ColorNatural
Fit for PurposePCR Ready
GraduatedYes
MaterialPolypropylene (Virgin)
Max. RCF18,000 x g
SterilityNon-sterile
Unit SizePack of 500