ART™ Reload System
Thermo Scientific™
ART™ Reload System
Reduce waste and save benchtop and storage space with this advanced nonfiltered reload system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)For Use With (Equipment)ModelFit for Purpose
3502-05-RTPK
also known as 3502-05-RT
10 μLUniversal FitART Non-filteredCertified Quality
3101-05-RTPK
also known as 3101-05-RT
1000 μLUniversal FitART Non-filteredSterile
3522-RTPK
also known as 3522-RT
20 μLEppendorfART Ultra Micro Non-FilteredCertified Quality
3102-05-RTPK
also known as 3102-05-RT
1000 μLUniversal FitART Non-filteredCertified Quality
3501-05-RTPK
also known as 3501-05-RT
10 μLUniversal FitART Non-filteredSterile
3501-RTPK10 μLUniversal FitART Non-filteredSterile
3521-05-RTPK
also known as 3521-05-RT
20 μLEppendorfART Non-filteredSterile
3521-RTPK
also known as 3521-RT
20 μLEppendorfART Non-filteredSterile
3522-05-RTPK
also known as 3522-05-RT
20 μLEppendorfART Non-filteredCertified Quality
3551-05-RTPK
also known as 3551-05-RT
200 μLUniversal FitART Non-filteredSterile
3771-05-RTPK
also known as 3771-05-RT
300 μLUniversal FitART Non-filteredSterile
3771-RTPK
also known as 3771-RT
300 μLUniversal FitART Non-filteredSterile
3772-05-RTPK
also known as 3772-05-RT
300 μLUniversal FitART Non-filteredCertified Quality
3772-RTPK
also known as 3772-RT
300 μLUniversal FitART Non-filteredCertified Quality
Catalog number 3502-05-RTPK
also known as 3502-05-RT
Price (INR)
-
Volume (Metric):
10 μL
For Use With (Equipment):
Universal Fit
Model:
ART Non-filtered
Fit for Purpose:
Certified Quality
Reduce waste and save benchtop and storage space with the advanced Thermo Scientific™ ART™ Nonfiltered Reload System. This system provides an environmentally responsible option without compromising sterility.
Features:
  • Stackable barrier tip packaging allows for tips to pass through offset holes in lower insert
  • Blister sleeve guide rails align tip inserts for smooth transfer into the rack
  • DNase, RNase, DNA, ATP, and Endotoxin Free
  • Outer sleeve protects tips from exterior contamination while refilling inserts
  • Tip packaging option reduces plastic waste
  • Rigid insert design provides reliable tip loading
  • Lot number and description are printed on every insert
Note:
Empty rack required for -RT parts
Specifications
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
Length (English)1.2 in.
Length (Metric)3.13 cm
ModelART Non-filtered
Quantity4800/Cs.
ColorNatural
FilteredNon-filtered
Fit for PurposeCertified Quality
For Use With (Equipment)Universal Fit
FormatRack
Product LineART
SterilityNon-sterile
Tip StyleLow Retention
Volume (Metric)10 μL
Unit SizePack of 960